(4-Bromphenyl)essigsäureethylester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Ethyl-(4-bromphenyl)acetat
M-Clarity™ Qualitätsstufe = MQ100
631353-25G
631353-5G
SIAL631353-25GEA
49
EUR
SIAL631353-25G
SIAL631353-5G
(4-Bromphenyl)essigsäureethylester, Sigma-Aldrich®
(4-Bromphenyl)essigsäureethylester
Formel:
C₁₀H₁₁BrO₂ |
MDL:
MFCD00016333 CAS-Nummer: 14062-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...