Bis(4-nitrophenyl)carbonat ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Bis(4-nitrophenyl) carbonate
L00290.06
L00290.14
L00290.06EA
43.7
EUR
L00290.06
L00290.14
Bis(4-nitrophenyl)carbonat ≥98%
Bis(4-nitrophenyl)carbonat
Purity: 98%
CAS No.: 5070-13-3
Molecular Formula: C13H8N2O7
Formula Weight: 304.21
Melting Point: 136-139°
Beilstein Reference: 1892897
Formel:
C₁₃H₈N₂O₇ Molecular Weight: 304.22 g/mol Schmelzpunkt: 136…139 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007322 CAS-Nummer: 5070-13-3 EINECS: 225-775-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...