(4-Nitrophenyl)essigsäureethylester ≥98%
Lieferant: Thermo Scientific
Synonyme:
Ethyl-(4-nitrophenyl)acetat
A11101.06
A11101.14
A11101.22
A11101.06EA
50.4
EUR
A11101.06
A11101.14
A11101.22
(4-Nitrophenyl)essigsäureethylester ≥98%
(4-Nitrophenyl)essigsäureethylester
Purity: 98%
CAS No.: 5445-26-1
Molecular Formula: C10H11NO4
Formula Weight: 209.20
Melting Point: 62-66°
Beilstein Reference: 2053241
Formel:
C₁₀H₁₁NO₄ Molecular Weight: 209,2 g/mol Schmelzpunkt: 62…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017046 CAS-Nummer: 5445-26-1 EINECS: 226-646-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...