(5-Brompentyl)benzol
Lieferant: FLUOROCHEM
Synonyme:
1-Brom-5-phenylpentan
008939-1G
008939-5G
008939-25G
FLUO008939-1GEA
57.9
EUR
FLUO008939-1G
FLUO008939-5G
FLUO008939-25G
(5-Brompentyl)benzol
(5-Bromopentyl)benzene
Formel:
C₁₁H₁₅Br Molecular Weight: 227,14 g/mol |
MDL:
MFCD01075177 CAS-Nummer: 14469-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...