(Brommethyl)cyclopropan

Lieferant: COMBI-BLOCKS

OS-7629-5G OS-7629-25G OS-7629-100G
COBBOS-7629-5GEA 26.2 EUR
COBBOS-7629-5G COBBOS-7629-25G COBBOS-7629-100G
(Brommethyl)cyclopropan
(Brommethyl)cyclopropan
Formel: C₄H₇Br
Molecular Weight: 135 g/mol
Siedepunkt: 105
Dichte: 1.41 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00001306
CAS-Nummer: 7051-34-5
EINECS: 230-331-8
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR