Bicin, Calbiochem®
Lieferant: Merck Millipore Calbiochem
Synonyme:
N,N-Bis(2-hydroxyethyl)glycin
391336-1
391336-100
391336-1EA
855
EUR
391336-1
391336-100
Bicin, Calbiochem®
Bicin
Formel:
C₆H₁₃NO₄ Molecular Weight: 163,17 g/mol Schmelzpunkt: 185…192 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004295 CAS-Nummer: 150-25-4 EINECS: 205-755-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...