(Methoxymethyl)triphenylphosphoniumchlorid

Lieferant: COMBI-BLOCKS
Achtung

OS-7557-500G OS-7557-25G OS-7557-100G
COBBOS-7557-500GEA 335 EUR
COBBOS-7557-500G COBBOS-7557-25G COBBOS-7557-100G
(Methoxymethyl)triphenylphosphoniumchlorid
(Methoxymethyl)triphenylphosphonium chloride
Formel: C₂₀H₂₀ClOP
Molecular Weight: 342,8 g/mol
Schmelzpunkt: 190 °C
MDL: MFCD00011800
CAS-Nummer: 4009-98-7
EINECS: 223-664-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR