(R)-(-)-Carvon 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: L(-)-Carvon, (R)-(-)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-on, L-p-Mentha-1(6),8-dien-2-on

154590050 154591000 154595000
ACRO154590050EA 21.2 EUR
ACRO154590050 ACRO154591000 ACRO154595000
(R)-(-)-Carvon 99%
(R)-(-)-Carvon
Formel: C₁₀H₁₄O
Molecular Weight: 150.22 g/mol
Siedepunkt: 227
Schmelzpunkt: 25,2
Dichte: 0.958 g/cm³
Flammpunkt: 88 °C
MDL: MFCD00001578
CAS-Nummer: 6485-40-1
EINECS: 229-352-5

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4950 to 1.5010 (20°C, 589 nm)
GC ≥98.5 %
Specific gravity (25/25°C) 0.956 to 0.961
Specific optical rotation -57° to -62° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR