D(+)-Tyrosin

Lieferant: COMBI-BLOCKS

Synonyme: (R)-(+)-p-Hydroxyphenylalanin, H-D-Tyr-OH, (R)-(+)-2-Amino-3-(4-hydroxyphenyl)propionsäure, D(+)-p-Hydroxyphenylalanin

OS-7696-5G OS-7696-25G OS-7696-100G
COBBOS-7696-5GEA 28.5 EUR
COBBOS-7696-5G COBBOS-7696-25G COBBOS-7696-100G
D(+)-Tyrosin
D(+)-Tyrosin
Formel: C₉H₁₁NO₃
Molecular Weight: 181.19 g/mol
Siedepunkt: 351
Schmelzpunkt: 285
MDL: MFCD00063073
CAS-Nummer: 556-02-5
EINECS: 209-112-6
Merck Index: 12,09970

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR