1,4-Dithioerythrit
Lieferant: BACHEM BIOCHEMICA
|
Achtung
|
Synonyme:
(R*,S*)-1,4-Dimercaptobutan-2,3-diol, erythro-1,4-Dimercaptobutan-2,3-diol, DTE, Dithioerythritol
4011255.0025
4011255.0100
BACE4011255.0025EA
0
EUR
BACE4011255.0025
BACE4011255.0100
1,4-Dithioerythrit
1,4-Dithioerythrit
Formel:
C₄H₁₀O₂S₂ Molecular Weight: 154.25 g/mol Siedepunkt: 286 Schmelzpunkt: 82…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063750 CAS-Nummer: 6892-68-8 EINECS: 229-998-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...