(S)-(+)-2-Amino-3-methyl-1-butanol
Lieferant: COMBI-BLOCKS
Synonyme:
L(+)-Valinol, L(+)-2-Amino-3-methyl-1-butanol
HC-6495-5G
HC-6495-25G
HC-6495-1G
HC-6495-100G
COBBHC-6495-5GEA
28.6
EUR
COBBHC-6495-5G
COBBHC-6495-25G
COBBHC-6495-1G
COBBHC-6495-100G
(S)-(+)-2-Amino-3-methyl-1-butanol
(S)-(+)-2-Amino-3-methyl-1-butanol
Formel:
C₅H₁₃NO Molecular Weight: 103,16 g/mol Siedepunkt: 187 °C (1013 hPa) Schmelzpunkt: 31…32 °C Dichte: 0,92 g/cm³ (20 °C) |
MDL:
MFCD00064296 CAS-Nummer: 2026-48-4 EINECS: 217-975-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...