(S)-(+)-Carvon (natürlich) 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: D(+)-Carvon, (S)-(+)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-on, D-p-Mentha-1(6),8-dien-2-on

150680250 150681000
ACRO150680250EA 32.3 EUR
ACRO150680250 ACRO150681000
(S)-(+)-Carvon (natürlich) 96%
(S)-(+)-Carvon
Formel: C₁₀H₁₄O
Molecular Weight: 150,22 g/mol
Siedepunkt: 230 °C (1013 hPa)
Schmelzpunkt: ? 15 °C
Dichte: 0,956 g/cm³
Flammpunkt: 97 °C
MDL: MFCD00062997
CAS-Nummer: 2244-16-8
EINECS: 218-827-2

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4960 to 1.5010 (20°C, 589 nm)
GC ≥95 %
Specific gravity (25°C) 0.953 to 0.962
Specific optical rotation +52.0° to +62.0° (20°C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR