(S)-(+)-Carvon 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: D(+)-Carvon, (S)-(+)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-on, D-p-Mentha-1(6),8-dien-2-on

L07130.14 L07130.22
L07130.14EA 43 EUR
L07130.14 L07130.22
(S)-(+)-Carvon 96%
(S)-(+)-Carvon

Purity: 96% CAS No.: 2244-16-8 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 93°(199°F) Refractive Index: 1.4980 Odour: Pleasant Boiling Point: 228-230° Beilstein Reference: 2042970 Merck Reference: 14,1874 Optical Rotation: +59° (neat) MDL No.: MFCD00062997

Formel: C₁₀H₁₄O
Siedepunkt: 228…230 °C
Schmelzpunkt: ? 15 °C
Dichte: 0,959
Flammpunkt: 93 °C (199 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00062997
CAS-Nummer: 2244-16-8
EINECS: 218-827-2
Merck Index: 14,1874

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR