Tyramin Hydrochlorid 98%
Lieferant: Thermo Scientific
Achtung
Synonyme: 2-(4-Hydroxyphenyl)ethylammoniumchlorid, 4-Hydroxyphenethylammoniumchlorid, 4-(2-Aminoethyl)phenol Hydrochlorid
A12220.06 A12220.14
A12220.06EA 29 EUR
A12220.06 A12220.14
Tyramin Hydrochlorid 98%
Tyramin Hydrochlorid
Purity: 98% CAS No.: 60-19-5 Molecular Formula: C8H11NO.HCl Formula Weight: 173.65 Melting Point: 271-274° Beilstein Reference: 3627058 Merck Reference: 14,9835 MDL No.: MFCD00012901
Formel: C₈H₁₂ClNO
Molecular Weight: 173,64 g/mol
Schmelzpunkt: 271…274 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012901
CAS-Nummer: 60-19-5
EINECS: 200-462-5
Merck Index: 14,9835