1-(2-Bromethoxy)-2-nitrobenzol

Lieferant: COMBI-BLOCKS

AN-1136-5G AN-1136-25G AN-1136-100G
COBBAN-1136-5GEA 69 EUR
COBBAN-1136-5G COBBAN-1136-25G COBBAN-1136-100G
1-(2-Bromethoxy)-2-nitrobenzol
1-(2-Bromethoxy)-2-nitrobenzol
Formel: C₈H₈BrNO₃
Molecular Weight: 246,06 g/mol
MDL: MFCD02030589
CAS-Nummer: 18800-37-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR