1-(2-Bromethoxy)-3-nitrobenzol

Lieferant: COMBI-BLOCKS

AN-1135-25G AN-1135-5G AN-1135-1G
COBBAN-1135-25GEA 204 EUR
COBBAN-1135-25G COBBAN-1135-5G COBBAN-1135-1G
1-(2-Bromethoxy)-3-nitrobenzol
1-(2-Bromethoxy)-3-nitrobenzol
Formel: C₈H₈BrNO₃
Molecular Weight: 246,06 g/mol
MDL: MFCD07783648
CAS-Nummer: 13831-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR