1-(2-Bromphenyl)-3,5-dimethyl-1H-pyrazol
Lieferant: COMBI-BLOCKS
Synonyme:
1-(2-Bromphenyl)-3,5-dimethyl-1H-pyrazol
HC-2982-5G
HC-2982-1G
COBBHC-2982-5GEA
375
EUR
COBBHC-2982-5G
COBBHC-2982-1G
1-(2-Bromphenyl)-3,5-dimethyl-1H-pyrazol
1-(2-Bromophenyl)-3,5-dimethyl-1H-pyrazole
Formel:
C₁₁H₁₁BrN₂ |
MDL:
MFCD12134495 CAS-Nummer: 1155573-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...