1-(3-Bromphenyl)piperazin

Lieferant: COMBI-BLOCKS

AM-1739-5G AM-1739-1G
COBBAM-1739-5GEA 459 EUR
COBBAM-1739-5G COBBAM-1739-1G
1-(3-Bromphenyl)piperazin
1-(3-Bromphenyl)piperazin
Formel: C₁₀H₁₃BrN₂
Molecular Weight: 241,13 g/mol
MDL: MFCD03412123
CAS-Nummer: 31197-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR