1-(3-Bromphenyl)-3,5-dimethyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

Synonyme: 1-(3-Bromphenyl)-3,5-dimethyl-1H-pyrazol

HC-2981-1G HC-2981-5G
COBBHC-2981-1GEA 216 EUR
COBBHC-2981-1G COBBHC-2981-5G
1-(3-Bromphenyl)-3,5-dimethyl-1H-pyrazol
1-(3-Bromophenyl)-3,5-dimethyl-1H-pyrazole
Formel: C₁₁H₁₁BrN₂
MDL: MFCD11845717
CAS-Nummer: 294877-29-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR