1-(3-Fluorphenyl)imidazol
Lieferant: FLUOROCHEM
Synonyme:
1-(3-Fluorphenyl)-1H-imidazol
002570-1G
002570-5G
FLUO002570-1GEA
87.9
EUR
FLUO002570-1G
FLUO002570-5G
1-(3-Fluorphenyl)imidazol
1-(3-Fluorophenyl)imidazole
Formel:
C₉H₇FN₂ |
MDL:
MFCD00060486 CAS-Nummer: 25372-42-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...