1-(4-Bromphenyl)piperazin

Lieferant: COMBI-BLOCKS

OS-7566-5G OS-7566-25G OS-7566-1G
COBBOS-7566-5GEA 125 EUR
COBBOS-7566-5G COBBOS-7566-25G COBBOS-7566-1G
1-(4-Bromphenyl)piperazin
1-(4-Bromphenyl)piperazin
Formel: C₁₀H₁₃BrN₂
Molecular Weight: 241,13 g/mol
MDL: MFCD00130198
CAS-Nummer: 66698-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR