1-(4-Bromphenyl)-3,5-dimethyl-1H-pyrazol

Lieferant: COMBI-BLOCKS

Synonyme: 1-(4-Bromphenyl)-3,5-dimethyl-1H-pyrazol

HC-2980-5G HC-2980-1G
COBBHC-2980-5GEA 451 EUR
COBBHC-2980-5G COBBHC-2980-1G
1-(4-Bromphenyl)-3,5-dimethyl-1H-pyrazol
1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole
Formel: C₁₁H₁₁BrN₂
MDL: MFCD09817558
CAS-Nummer: 62546-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR