1-(4-Bromphenyl)imidazol

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Bromphenyl)-1H-imidazol

018307-1G 018307-20G
FLUO018307-1GEA 130 EUR
FLUO018307-1G FLUO018307-20G
1-(4-Bromphenyl)imidazol
1-(4-Bromophenyl)imidazole
Formel: C₉H₇BrN₂
Molecular Weight: 223.07 g/mol
MDL: MFCD00060489
CAS-Nummer: 10040-96-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR