1-(4-Fluorphenyl)imidazol

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Fluorphenyl)-1H-imidazol

002571-1G 002571-5G
FLUO002571-1GEA 155 EUR
FLUO002571-1G FLUO002571-5G
1-(4-Fluorphenyl)imidazol
1-(4-Fluorophenyl)imidazole
Formel: C₉H₇FN₂
Molecular Weight: 162.17 g/mol
MDL: MFCD00060490
CAS-Nummer: 21441-24-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR