ω-Brom-3,4-dichloracetophenon

Lieferant: Thermo Scientific

Synonyme: α-Brom-3,4-dichloracetophenon, 2-Brom-1-(3,4-dichlorphenyl)ethan-1-on, 3,4-Dichlorphenacylbromid, 2-Brom-3',4'-dichloracetophenon

BTB02913DA BTB02913EA
MAYBBTB02913DAEA 0 EUR
MAYBBTB02913DA MAYBBTB02913EA
ω-Brom-3,4-dichloracetophenon
ω-Brom-3,4-dichloracetophenon
Formel: C₈H₅BrCl₂O
Molecular Weight: 267,94 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00051581
CAS-Nummer: 2632-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR