ω-Chlor-2,4-difluoracetophenon
Lieferant: COMBI-BLOCKS
Synonyme:
2-Chlor-2',4'-difluoracetophenon, α-Chlor-2,4-difluoracetophenon
CA-4154-5G
CA-4154-25G
CA-4154-100G
COBBCA-4154-5GEA
51.3
EUR
COBBCA-4154-5G
COBBCA-4154-25G
COBBCA-4154-100G
ω-Chlor-2,4-difluoracetophenon
ω-Chlor-2,4-difluoracetophenon
Formel:
C₈H₅ClF₂O Molecular Weight: 190,58 g/mol Siedepunkt: 233 °C (1013 hPa) Schmelzpunkt: 47…48 °C |
MDL:
MFCD00013252 CAS-Nummer: 51336-94-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...