α'-Chlor-α,α,α-trifluor-o-xylol

Lieferant: FLUOROCHEM

Synonyme: 2-(Chlormethyl)benzotrifluorid, 2-(Trifluormethyl)benzylchlorid, 1-(Chlormethyl)-2-(trifluormethyl)benzol

007608-25G 007608-5G
FLUO007608-25GEA 112 EUR
FLUO007608-25G FLUO007608-5G
α'-Chlor-α,α,α-trifluor-o-xylol
α'-Chlor-α,α,α-trifluor-o-xylol
Formel: C₈H₆ClF₃
Molecular Weight: 194,58 g/mol
Siedepunkt: 103…110 °C (25 mmHg)
Dichte: 1,3079 g/cm³ (20 °C)
MDL: MFCD00009920
CAS-Nummer: 21742-00-7
EINECS: 244-560-6
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR