α-Chlor-4-(trifluormethoxy)toluol

Lieferant: FLUOROCHEM

Synonyme: 4-(Trifluormethoxy)benzylchlorid, p-(Trifluormethoxy)benzylchlorid

007553-1G 007553-5G
FLUO007553-1GEA 69.8 EUR
FLUO007553-1G FLUO007553-5G
α-Chlor-4-(trifluormethoxy)toluol
α-Chlor-4-(trifluormethoxy)toluol
Formel: C₈H₆ClF₃O
Molecular Weight: 210,58 g/mol
MDL: MFCD00052326
CAS-Nummer: 65796-00-1
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR