α'-Chlor-α,α,α-trifluor-p-xylol

Lieferant: FLUOROCHEM

Synonyme: 4-(Trifluormethyl)benzylchlorid, 4-(Chlormethyl)benzotrifluorid, 1-(Chlormethyl)-4-(trifluormethyl)benzol

007610-5G 007610-25G
FLUO007610-5GEA 36.9 EUR
FLUO007610-5G FLUO007610-25G
α'-Chlor-α,α,α-trifluor-p-xylol
α'-Chlor-α,α,α-trifluor-p-xylol
Formel: C₈H₆ClF₃
Molecular Weight: 194.58 g/mol
Siedepunkt: 47…49 °C (20 mmHg)
Schmelzpunkt: 19
Dichte: 1.31 g/cm³ (20 °C)
MDL: MFCD00040772
CAS-Nummer: 939-99-1
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR