2-(Trifluormethyl)phenylacetonitril
Lieferant: COMBI-BLOCKS
Synonyme:
2-(Trifluormethyl)benzylcyanid
OS-7611-5G
OS-7611-25G
OS-7611-100G
COBBOS-7611-5GEA
50.8
EUR
COBBOS-7611-5G
COBBOS-7611-25G
COBBOS-7611-100G
2-(Trifluormethyl)phenylacetonitril
2-(Trifluormethyl)phenylacetonitril
Formel:
C₉H₆F₃N Molecular Weight: 185,15 g/mol Siedepunkt: 113…116 °C (23 mmHg) Schmelzpunkt: 29…30 °C |
MDL:
MFCD00001903 CAS-Nummer: 3038-47-9 EINECS: 221-239-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...