3-(Trifluormethoxy)phenylacetonitril
Lieferant: FLUOROCHEM
Synonyme:
3-(Trifluormethoxy)benzylcyanid
007565-1G
007565-5G
FLUO007565-1GEA
162
EUR
FLUO007565-1G
FLUO007565-5G
3-(Trifluormethoxy)phenylacetonitril
3-(Trifluormethoxy)phenylacetonitril
Formel:
C₉H₆F₃NO Molecular Weight: 201,15 g/mol |
MDL:
MFCD00061272 CAS-Nummer: 108307-56-8 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...