3-(Trifluormethyl)phenylacetonitril

Lieferant: FLUOROCHEM

Synonyme: 3-(Trifluormethyl)benzylcyanid

007636-10G 007636-50G 007636-250G
FLUO007636-10GEA 30.8 EUR
FLUO007636-10G FLUO007636-50G FLUO007636-250G
3-(Trifluormethyl)phenylacetonitril
3-(Trifluormethyl)phenylacetonitril
Formel: C₉H₆F₃N
Molecular Weight: 185.15 g/mol
Siedepunkt: 92
Dichte: 1.18
MDL: MFCD00001913
CAS-Nummer: 2338-76-3
EINECS: 219-053-8
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR