2-Hydroxy-3'-nitroacetophenon

Lieferant: BIONET RESEARCH

Synonyme: α-Hydroxy-3'-nitroacetophenon

3X-0728.500 3X-0728.1 3X-0728.5 3X-0728.10
BIOT3X-0728.500EA 0 EUR
BIOT3X-0728.500 BIOT3X-0728.1 BIOT3X-0728.5 BIOT3X-0728.10
2-Hydroxy-3'-nitroacetophenon
2-Hydroxy-3'-nitroacetophenon
Formel: C₈H₇NO₄
Molecular Weight: 181.15
MDL: MFCD04973723
CAS-Nummer: 72802-41-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR