2-Hydroxy-4'-methoxyacetophenon
Lieferant: BIONET RESEARCH
Synonyme:
α-Hydroxy-4'-methoxyacetophenon
4X-0724.500
4X-0724.1
4X-0724.5
4X-0724.10
BIOT4X-0724.500EA
0
EUR
BIOT4X-0724.500
BIOT4X-0724.1
BIOT4X-0724.5
BIOT4X-0724.10
2-Hydroxy-4'-methoxyacetophenon
2-Hydroxy-4'-methoxyacetophenon
Formel:
C₉H₁₀O₃ Molecular Weight: 166,18 g/mol |
MDL:
MFCD00004668 CAS-Nummer: 4136-21-4 EINECS: 223-955-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...