2-Hydroxy-4'-nitroacetophenon

Lieferant: BIONET RESEARCH

Synonyme: α-Hydroxy-4'-nitroacetophenon

6X-0701.500 6X-0701.1 6X-0701.5 6X-0701.10
BIOT6X-0701.500EA 0 EUR
BIOT6X-0701.500 BIOT6X-0701.1 BIOT6X-0701.5 BIOT6X-0701.10
2-Hydroxy-4'-nitroacetophenon
2-Hydroxy-4'-nitroacetophenon
Formel: C₈H₇NO₄
Molecular Weight: 181,15 g/mol
MDL: MFCD00599385
CAS-Nummer: 64611-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR