3,4,5-Trifluorbenzophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(Phenylmethanoyl)-3,4,5-trifluorbenzene

007492-1G 007492-5G
FLUO007492-1GEA 39.9 EUR
FLUO007492-1G FLUO007492-5G
3,4,5-Trifluorbenzophenon
3,4,5-Trifluorbenzophenon
Formel: C₁₃H₇F₃O
Molecular Weight: 236.19 g/mol
MDL: MFCD00083525
CAS-Nummer: 70028-88-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR