4'-Methyl-2,2,2-trifluoracetophenon

Lieferant: Thermo Scientific

Synonyme: 2,2,2-Trifluor-1-(4-methylphenyl)-1-ethanon

TL00424.1 TL00424.10 TL00424.50
MAYBTL00424.1EA 0 EUR
MAYBTL00424.1 MAYBTL00424.10 MAYBTL00424.50
4'-Methyl-2,2,2-trifluoracetophenon
4'-Methyl-2,2,2-trifluoracetophenon
Formel: C₉H₇F₃O
Molecular Weight: 188.15 g/mol
MDL: MFCD00052338
CAS-Nummer: 394-59-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR