4-(Trifluormethoxy)benzotrifluorid

Lieferant: FLUOROCHEM

Synonyme: 1-(Trifluormethoxy)-4-(trifluormethyl)benzol

007977-5G 007977-1G
FLUO007977-5GEA 112 EUR
FLUO007977-5G FLUO007977-1G
4-(Trifluormethoxy)benzotrifluorid
4-(Trifluormethoxy)benzotrifluorid
Formel: C₈H₄F₆O
Molecular Weight: 230.11 g/mol
MDL: MFCD00190125
CAS-Nummer: 80258-33-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR