2,3-Diaminophenol

Lieferant: COMBI-BLOCKS

Synonyme: 3-Hydroxy-o-phenylendiamin

AN-1450-1G AN-1450-5G AN-1450-25G
COBBAN-1450-1GEA 0 EUR
COBBAN-1450-1G COBBAN-1450-5G COBBAN-1450-25G
2,3-Diaminophenol
2,3-Diaminophenol
Formel: C₆H₈N₂O
MDL: MFCD00075199
CAS-Nummer: 59649-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR