4-tert-Butyl-o-phenylendiamin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1,2-Diamino-4-tert-butylbenzol, 4-tert-Butylbenzol-1,2-diamin, 4-tert-Butyl-1,2-phenylendiamin

H32205.PG H32205.03 H32205.06
H32205.PGEA 0 EUR
H32205.PG H32205.03 H32205.06
4-tert-Butyl-o-phenylendiamin 97%
4-tert-Butyl-o-phenylendiamin

Purity: 97% CAS No.: 68176-57-8 Molecular Formula: C10H16N2 Formula Weight: 164.25 Melting Point: 98-100° UN No.: 2811 MDL No.: MFCD00052695

Formel: C₁₀H₁₆N₂
Schmelzpunkt: 98…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052695
CAS-Nummer: 68176-57-8
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR