4-tert-Butyl-o-phenylendiamin

Lieferant: FLUOROCHEM

Synonyme: 1,2-Diamino-4-tert-butylbenzol, 4-tert-Butylbenzol-1,2-diamin, 4-tert-Butyl-1,2-phenylendiamin

017188-1G 017188-5G
FLUO017188-1GEA 44.4 EUR
FLUO017188-1G FLUO017188-5G
4-tert-Butyl-o-phenylendiamin
4-tert-Butyl-o-phenylendiamin
Formel: C₁₀H₁₆N₂
Molecular Weight: 164,25 g/mol
MDL: MFCD00052695
CAS-Nummer: 68176-57-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR