4,5-Dichlor-o-phenylendiamin

Lieferant: COMBI-BLOCKS

Synonyme: 1,2-Diamino-4,5-dichlorbenzol, 4,5-Dichlor-1,2-phenylendiamin, 4,5-Dichlorbenzol-1,2-diamin

HC-6207-5G HC-6207-25G HC-6207-1G
COBBHC-6207-5GEA 50.9 EUR
COBBHC-6207-5G COBBHC-6207-25G COBBHC-6207-1G
4,5-Dichlor-o-phenylendiamin
4,5-Dichlor-o-phenylendiamin
Formel: C₆H₆Cl₂N₂
Molecular Weight: 177.03 g/mol
Siedepunkt: 305
Schmelzpunkt: 163
MDL: MFCD00007723
CAS-Nummer: 5348-42-5
EINECS: 226-305-0
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR