3',4'-Dichloracetophenon

Lieferant: Thermo Scientific

Synonyme: 1-(3,4-dichlorophenyl)ethanone

BTB04486.1 BTB04486.10 BTB04486.50
MAYBBTB04486.1EA 0 EUR
MAYBBTB04486.1 MAYBBTB04486.10 MAYBBTB04486.50
3',4'-Dichloracetophenon
3',4'-Dichloracetophenon
Formel: C₈H₆Cl₂O
Molecular Weight: 189,04 g/mol
Siedepunkt: 265 °C (1013 hPa)
Schmelzpunkt: 44…46 °C
MDL: MFCD00000553
CAS-Nummer: 2642-63-9
EINECS: 220-146-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR