3,4-Dichlor-6-nitrotoluol
Lieferant: COMBI-BLOCKS
Synonyme:
4,5-Dichloro-2-nitrotoluene, 1,2-Dichloro-4-methyl-5-nitrobenzene
AN-1417-5G
AN-1417-25G
AN-1417-1G
COBBAN-1417-5GEA
137
EUR
COBBAN-1417-5G
COBBAN-1417-25G
COBBAN-1417-1G
3,4-Dichlor-6-nitrotoluol
3,4-Dichlor-6-nitrotoluol
Formel:
C₇H₅Cl₂NO₂ Molecular Weight: 206.03 g/mol |
MDL:
MFCD01109847 CAS-Nummer: 7494-45-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...