2',3'-Difluor-4'-methylacetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(2,3-Difluoro-4-methylphenyl)ethanone
005742-1G
005742-5G
FLUO005742-1GEA
87.9
EUR
FLUO005742-1G
FLUO005742-5G
2',3'-Difluor-4'-methylacetophenon
2',3'-Difluor-4'-methylacetophenon
Formel:
C₉H₈F₂O Molecular Weight: 170.16 g/mol |
MDL:
MFCD01631650 CAS-Nummer: 261763-30-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...