2,3-Difluor-6-nitrophenol
Lieferant: COMBI-BLOCKS
Synonyme:
1,2-Difluoro-3-hydroxy-4-nitrobenzene
AN-1358-100G
AN-1358-25G
AN-1358-5G
COBBAN-1358-100GEA
0
EUR
COBBAN-1358-100G
COBBAN-1358-25G
COBBAN-1358-5G
2,3-Difluor-6-nitrophenol
2,3-Difluor-6-nitrophenol
Formel:
C₆H₃F₂NO₃ Schmelzpunkt: 61 °C |
MDL:
MFCD00042255 CAS-Nummer: 82419-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...