2,3-Difluor-4-methoxyphenol

Lieferant: FLUOROCHEM

Synonyme: 1,2-Difluor-3-hydroxy-6-methoxybenzol, 2,3-Difluor-4-hydroxyanisol

005741-1G 005741-5G
FLUO005741-1GEA 104 EUR
FLUO005741-1G FLUO005741-5G
2,3-Difluor-4-methoxyphenol
2,3-Difluor-4-methoxyphenol
Formel: C₇H₆F₂O₂
Molecular Weight: 160.12 g/mol
MDL: MFCD01631505
CAS-Nummer: 261763-29-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR