2,3-Difluorphenol

Lieferant: COMBI-BLOCKS

OS-7590-5G OS-7590-25G
COBBOS-7590-5GEA 38.7 EUR
COBBOS-7590-5G COBBOS-7590-25G
2,3-Difluorphenol
2,3-Difluorphenol
Formel: C₆H₄F₂O
Molecular Weight: 130.09 g/mol
Siedepunkt: 54 °C (25 mmHg)
Schmelzpunkt: 30…32 °C
MDL: MFCD00010262
CAS-Nummer: 6418-38-8
UN: 2921
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR