2,3-Difluor-4-nitroanisol

Lieferant: COMBI-BLOCKS

AN-1156-5G AN-1156-25G AN-1156-1G
COBBAN-1156-5GEA 76.4 EUR
COBBAN-1156-5G COBBAN-1156-25G COBBAN-1156-1G
2,3-Difluor-4-nitroanisol
2,3-Difluor-4-nitroanisol
Formel: C₇H₅F₂NO₃
Molecular Weight: 189,12 g/mol
MDL: MFCD07995055
CAS-Nummer: 66684-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR