3',4'-Dimethoxyacetophenon

Lieferant: Thermo Scientific

Synonyme: 1-(3,4-Dimethoxyphenyl)ethan-1-on

SB01100.1 SB01100.10 SB01100.50
MAYBSB01100.1EA 0 EUR
MAYBSB01100.1 MAYBSB01100.10 MAYBSB01100.50
3',4'-Dimethoxyacetophenon
3',4'-Dimethoxyacetophenon
Formel: C₁₀H₁₂O₃
Molecular Weight: 180.2 g/mol
Siedepunkt: 286
Schmelzpunkt: 48…49 °C
MDL: MFCD00008737
CAS-Nummer: 1131-62-0
EINECS: 214-468-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR